missing translation for 'onlineSavingsMsg'
Learn More

Twenty two elements solution for ICP 100mg/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical MS 103 500


 View more versions of this product

Product Code. 12996213

  • 560.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Twenty two elements solution - As, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb Sb Se Sr, Ti, Tl, V, Zn for ICP 100mg/L HNO3 matrix
Specifications

Specifications

Twenty-Two Elements Solution
Bottle
ICP Multi-element Standard Solution
As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V, Zn
Liquid
100mg/L
500 mL
1760
5% HNO3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Multi-element Standard Solution for ICP. 22 Components in HNO3 5% (MS 103)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH207-Warning

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers