missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Twenty two elements solution for ICP 100mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
317.00 € - 560.00 €
Specifications
Chemical Name or Material | Twenty-Two Elements Solution |
---|---|
Concentration | 100mg/L |
Packaging | Bottle |
Type | ICP Multi-element Standard Solution |
UN Number | 1760 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12976213
|
Fisher Chemical
MS 103 050 |
50 mL | Bottle |
390.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12986213
|
Fisher Chemical
MS 103 100 |
100 mL | Bottle |
317.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12996213
|
Fisher Chemical
MS 103 500 |
500 mL | Bottle |
560.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Twenty two elements solution - As, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb Sb Se Sr, Ti, Tl, V, Zn for ICP 100mg/L HNO3 matrixSpecifications
Twenty-Two Elements Solution | |
Bottle | |
1760 | |
5% HNO3 |
100mg/L | |
ICP Multi-element Standard Solution | |
As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V, Zn | |
Liquid |
Safety and Handling
H314 ; H317 ; H332 ; H350 ; H412
P202;P260;P264;P271;P272;P273;P280;P301+P330+P331;P303+P361+P353;P304 +P340;P305+P351+P338;P308+P313;P310;P333+P313;P362+P364;P501
Danger
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Multi-element Standard Solution for ICP. 22 Components in HNO3 5% (MS 103)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .