Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased



Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
Brand: Eppendorf™ 0030531043
151.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)

Specifications
Custom Barcoding Available | |
Eppendorf | |
BioBased PCR Plates | |
Polypropylene, Polycarbonate | |
Non-sterile | |
25 Pk. |
8 x 12 | |
Colorless | |
Semi-skirted | |
96 | |
Conical Bottom (V-well) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.