Learn More
Lenz Laborglasintrumente™ Two-Neck Round-Bottom Flasks
Distillation flasks with parallel side neck and conical ground joints
Brand:  Lenz Laborglasintrumente™ 03204149
 View more versions of this product
Product Code. 10240881
  • 34.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
Center neck - NS 29/32, Side Neck - NS 14/23
Borosilicate Glass (3.3)
85 mm
250 mL
Distillation