missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente™ Two-Neck Round-Bottom Flasks
Distillation flasks with parallel side neck and conical ground joints
34.00 € - 46.00 €
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Distillation |
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
10240881
|
Lenz Laborglasintrumente™
03204149 |
250 mL | Center neck - NS 29/32, Side Neck - NS 14/23 |
34.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
10645231
|
Lenz Laborglasintrumente™
03204449 |
250 mL | Center neck - NS 29/32, Side Neck - NS 29/32 |
40.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
10560132
|
Lenz Laborglasintrumente™
03204458 |
500 mL | Center neck - NS 29/32, Side Neck - NS 29/32 |
46.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
Description
- With inscription label
- Material: DURAN™
Specifications
Borosilicate Glass (3.3) | |
Distillation |