Learn More
Lenz Laborglasinstrumente™ Two-Neck Round-Bottom Flasks, Side Neck Angled
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
37.00 € - 135.00 €
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
10519951
|
Lenz Laborglasinstrumente™
03214173 |
2 L | Side Neck NS 14/23 |
134.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10752682
|
Lenz Laborglasinstrumente™
03211149 |
250 mL | Side Neck NS 14/23 |
53.00 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
10230881
|
Lenz Laborglasinstrumente™
03214470 |
1 L | Side Neck 29/32 |
58.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10439362
|
Lenz Laborglasinstrumente™
03214273 |
2 L | Side Neck NS 19/26 |
135.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10010251
|
Lenz Laborglasinstrumente™
03213258 |
500 mL | Side Neck NS 19/26 |
61.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10384971
|
Lenz Laborglasinstrumente™
03213237 |
100 mL | Side Neck NS 19/26 |
41.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10394971
|
Lenz Laborglasinstrumente™
03213249 |
250 mL | Side Neck NS 19/26 |
55.00 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
10479932
|
Lenz Laborglasinstrumente™
03214473 |
2 L | Side Neck 29/32 |
134.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
10728271
|
Lenz Laborglasinstrumente™
03213137 |
100 mL | Side Neck NS 14/23 |
37.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
Description
- Distillation flasks acc. to DIN 12394 and in addition to DIN standard
- With NS
- With angled side neck (20°)
- With inscription label
- Material: DURAN™
Specifications
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Two-Neck Round-Bottom Flasks, Side Neck Angled