Learn More
Thermo Scientific™ TX-150 Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Perform a a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor, which is highly configurable and convenient.
Brand: Thermo Scientific™ 75005701
1032.48 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Perform a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor. This rotor accommodates up to 24 x 5/7mL blood collection tubes or 8 x 50mL conical tubes at 4,500rpm/ 3,260 x g, and provides additional flexibility for swing-out processing needs with available adapters. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ ClickSeal™ biocontainment lids—shorten run set-up time while providing peace-of-mind that samples are secure.
Designed for a broad range of swing-out processing needs at high speed and high capacity with dedicated buckets available for 8 conical tubes capacity or round buckets with ClickSeal biocontainment lids certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
- Trouble-free rotor installation and removal
- No tools are required
- No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Round buckets (75005702), 50mL Conical buckets (75005703) ClickSeal biocontainment lids (75005707) and replacement O-rings (75005724) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Specifications
TX-150 Swinging Bucket Rotor | |
4 x 145 mL | |
4,500 rpm | |
12968 | |
50 mm | |
90° |
Swinging Bucket Rotor | |
3,260 x g | |
144 mm | |
100 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges | |
Yes |