Learn More
Thermo Scientific™ TX-400 4 x 400 mL Swinging Bucket Rotor
Support a wide range of sample sizes and applications with the swinging bucket rotor.
Brand: Thermo Scientific™ 75003655
Description
Support a wide range of sample sizes and applications with the Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor.
Ordering Information:
Buckets and lids are sold separately; order rotor body (75-003-629) and components to complete assembly. For replacement O-rings for lids (set of four), order 75-003-657. Tube adapters are also available separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
Round Buckets for TX-400 Rotor (4) | |
4 x 400 mL | |
Biocontainment certified by CAMR™, HPA, Porton Down, UK |
Swinging Bucket Rotor | |
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.