missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Orange Hooded Coverall

Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Orange Hooded Coverall are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
928.00 €
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | Tychem |
Color | Orange |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12633680
![]() |
DuPont™
D13495222 |
2X-Large |
928.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12643680
![]() |
DuPont™
D13495244 |
Large |
928.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12653680
![]() |
DuPont™
D13495252 |
Medium |
928.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12663680
![]() |
DuPont™
D13495277 |
X-Large |
928.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
Description
- Available in grey for discretionary needs
- Elasticated face, wrists, waist and ankles for optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face masks
- Self-adhesive double zipper flap closure system
- Stitched and over-taped seams with barrier tape for protection and strength
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications
Hooded Coveralls | |
Orange | |
Elastic | |
Hooded | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
Self-adhesive chin flap | |
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications |
Tychem | |
Elastic | |
Elastic | |
Elastic | |
Stitched, Taped | |
Chemicals | |
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tychem™ 6000 F Orange Hooded Coverall