missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ NT450
Flexible impermeable knit-lined dual nitrile glove. Ideal for heavy duty work or longer exposure to hazardous chemicals and solvents.
73.80 €
Specifications
Certifications/Compliance (Europe) | CE Cat III |
---|---|
Color | Blue |
Exterior Finish | Rough |
Latex Free | No |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16370466
|
DuPont™
D15536299 |
Small, 8 | N/A | ||||||
16380466
|
DuPont™
D15536300 |
Medium, 9 |
73.80 €
12 pairs |
In Stock
Log in to see stock. |
|||||
16390466
|
DuPont™
D15536301 |
Large, 10 |
73.80 €
12 pairs |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
16300476
|
DuPont™
D15536302 |
X-Large, 11 | N/A | ||||||
Description
- Double nitrile coated gloves for jobs requiring chemical performance and comfort, plus excellent adherence
- Lightweight,'Second-Skin' feel and low-modulus formulation reduces hand fatigue
- CE CAT III - EN388: 4112X, EN ISO 374-1:2016/TYPE A, JKLOPT
- EN ISO 374-5:2016
- Featuring a cotton/polyester liner that absorbs perspiration for added comfort and a rough textured finish that ensures excellent grip
- Food contact compatible
- Resists Hydrochloric Acid (37%) - Level 6 (breakthrough time EN374)
Specifications
CE Cat III | |
Rough | |
Nitrile | |
56 mil | |
EN388: 4112X | |
Type A | |
Yes | |
Cotton/polyester liner | |
NT450 | |
Resists Hydrochloric Acid (37%) - Level 6 (breakthrough time EN374) | |
356 mm |
Blue | |
No | |
Scalloped edge / Anatomical shape | |
1.42 mm | |
No | |
JKLOPT | |
Plain | |
Lined | |
12 pairs per bag/6 bags per box: 72 total pairs | |
14 in. |
- Store away from light and humidity
- Rinse gloves in running water before removing, using a neutral detergent if necessary
- Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
- For Professional Use Only