Learn More
MedChemExpress Tyr-Gly-Gly-Phe-Met-OH
MedChemExpress Tyr-Gly-Gly-Phe-Met-OH regulates human immune function and inhibits tumor growth via binding to the opioid receptor.
Brand: MedChemExpress HY-P0073

Specifications
98.07% | |
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month | |
573.66 g/mol | |
25 mg | |
DMSO : ≥ 40 mg/mL (69.73 mM) ∣H2O : 6.67 mg/mL (11.63 mM∣Need ultrasonic) |
58569-55-4 | |
Cancer-programmed cell death | |
Peptides | |
Room temperature | |
Met-Enkephalin∣ Methionine enkephalin |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.