missing translation for 'onlineSavingsMsg'
Learn More

Tyrosinase Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tyrosinase Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-800-P0

 View more versions of this product

Product Code. 12642337

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients.

Host Species: Mouse

Clone: T311

Isotype: IgG2a

Species Reactivity: Human, Dog and Cat. Weakly cross-reacts with Mouse. Others not known.

Epitope: aa 1-433

Immunogen: Recombinant tyrosinase protein

Molecular Weight: 70-80kDa

Positive Control: Melanoma cell lines and Melanoma

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Staining of melanomas with Ab-1 showed tyrosinase in melanotic as well as amelanotic variants. Ab-1 is a useful marker for melanocytes and melanomas. Occasionally a minor band at 55kDa is also detected in Western Blotting

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunocytology
Specifications

Specifications

Tyrosinase Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Canine, Feline, Human
IgG2a
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
T311
Unconjugated
Recombinant tyrosinase protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.