Learn More
Tyrosinase Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Tyrosinase Ab-1, Mouse Monoclonal Antibody.
141.78 € - 875.00 €
Specifications
Antigen | Tyrosinase Ab-1 |
---|---|
Clone | T311 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12612337
|
Epredia
MS-800-B0 |
Concentrated | Biotin | 100 μL | N/A | ||||
12622337
|
Epredia
MS-800-B1 |
Concentrated | Biotin | 500 μL | N/A | ||||
12602337
|
Epredia
MS-800-B |
Concentrated | Biotin | 1 mL | N/A | ||||
12642337
|
Epredia
MS-800-P0 |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12662337
|
Epredia
MS-800-P1ABX |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12672337
|
Epredia
MS-800-PABX |
Concentrated | Unconjugated | 200 μL | N/A | ||||
12652337
|
Epredia
MS-800-P1 |
Concentrated | Unconjugated | 500 μL | N/A | ||||
12632337
|
Epredia
MS-800-P |
Concentrated | Unconjugated | 1 mL |
779.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12602347
|
Epredia
MS-800-R7 |
Prediluted | Unconjugated | 7 mL | N/A | ||||
12612347
|
Epredia
MS-800-RQ |
Ready-to-Use | Unconjugated | 12 mL | N/A | ||||
Description
Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients.Host Species: Mouse
Clone: T311
Isotype: IgG2a
Species Reactivity: Human, Dog and Cat. Weakly cross-reacts with Mouse. Others not known.
Epitope: aa 1-433
Immunogen: Recombinant tyrosinase protein
Molecular Weight: 70-80kDa
Positive Control: Melanoma cell lines and Melanoma
Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Staining of melanomas with Ab-1 showed tyrosinase in melanotic as well as amelanotic variants. Ab-1 is a useful marker for melanocytes and melanomas. Occasionally a minor band at 55kDa is also detected in Western Blotting
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunocytology
Specifications
Tyrosinase Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cancer and Tumor Biology | |
Primary |
T311 | |
Monoclonal | |
IVD | |
Recombinant tyrosinase protein |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.