Learn More
DuPont™ Tyvek™ IsoClean™ Coverall with Shoe Covers
Composed of flash spun high-density polyethylene. Dupont™ Tyvek™ IsoClean™ Coverall with Shoe Covers is permeable to both air and water vapor, yet repels water-based liquids and aerosols. Offers an excellent barrier against fine particles and fibers (down to 1μ in size) and is ultra-low-linting.
Brand: DuPont™ D14248016
This item is not returnable.
View return policy
Description
- Coverall with hood
- White
- Clean-processed and sterile
- Stitched internal seams
- Set sleeve design with elastication at wrists, ankles and face
- Attached thumb loops
- Zipper closure
- Attached overshoes with slip-retardant soles
- Not CE certified as chemical protective clothing
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2447150 | Certificate of Sterility | 17/12/2024 | D14248016 |
AHS2447144 | Certificate of Sterility | 17/12/2024 | D14248016 |
AHS2445129 | Certificate of Sterility | 11/12/2024 | D14248016 |
AHS2446124 | Certificate of Sterility | 11/12/2024 | D14248016 |
AHS2445172 | Certificate of Sterility | 11/12/2024 | D14248016 |
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Coverall with Shoe Covers >
Spot an opportunity for improvement?Share a Content Correction