Learn More
DuPont™ Tyvek™ IsoClean™ Slip-retardant Shoe Cover
Made of high density polyethylene, Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
146.00 €
Specifications
Color | White |
---|---|
Sterility | Non-sterile |
Model | IC451S00 |
Seam Type | Serged |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |
Description
- Gripper™ soles provide enhanced slip-retardance and robustness
- Delivers an ideal balance of protection, durability and comfort li>Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms
Specifications
White | |
IC451S00 | |
(EU) 2016/425, EN 14126 |
Non-sterile | |
Serged |