missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 2000 C Gown
Shin-length gown with wrap-over rear closure, hook and loop neck closure and waist ties offers enhanced protection for body parts that are more exposed to hazardous substances.
407.60 € - 432.48 €
Specifications
Product Type | General Purpose Gown |
---|---|
Gender | Unisex |
No. of Pockets | None |
Style | Wrap-around |
Material | Tychem |
Description
- Shin length
- Front protection with wrap-over rear closure
- Hook and loop neck closure and waist ties
- Stitched and over-taped seams
- Yellow
- Polymeric barrier coating provides permeation barrier protection against a wide range of inorganic chemicals and biological hazards
Specifications
General Purpose Gown | |
None | |
Tychem | |
Wrap-around Waist Ties | |
Non-sterile | |
Not Autoclavable |
Unisex | |
Wrap-around | |
Yellow | |
Single-use | |
Elastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title