missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Selfe™ U-Tube Reverse Flow Viscometers
Suitable for transparent and opaque liquids. With UKAS certificate of calibration
Brand: Poulten Selfe™ 1637/02
This item is not returnable.
View return policy
Description
Type BS/IP/RF, IP71, IP319, ASTM D2170.Suitable for transparent and opaque liquids. Nominal overall length 275mm. Approximate sample size 12mL to 25mL. With UKAS certificate of calibration.Specifications
U-Tube Reverse Flow Viscometers | |
D446 | |
Used for the measurement of kinematic viscosities of transparent and opaque. | |
12 to 25 mL |
2 to 20 cSt | |
UKAS | |
2 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Poulten Selfe™ U-Tube Reverse Flow Viscometers > Size: 2; Viscosity Range: 2 to 20mm2/s
Spot an opportunity for improvement?Share a Content Correction