Learn More
Poulten Selfe™ U-Tube Reverse Flow Viscometers
Suitable for transparent and opaque liquids. With UKAS certificate of calibration
Brand:  Poulten Selfe™ 1637/04
 View more versions of this product
Product Code. 13171033
  • 720.00 € / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Type BS/IP/RF, IP71, IP319, ASTM D2170.Suitable for transparent and opaque liquids. Nominal overall length 275mm. Approximate sample size 12mL to 25mL. With UKAS certificate of calibration.
Specifications
Specifications
U-Tube Reverse Flow Viscometers
D446
4
20 to 100 cSt
UKAS
12 to 25 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Poulten Selfe™ U-Tube Reverse Flow Viscometers > Size: 4; Viscosity Range: 20 to 100mm2/s