Learn More
Poulten Selfe™ U-Tube Viscometer
Suitable for transparent liquids. With UKAS certificate of calibration
227.00 € - 239.00 €
Specifications
Product Type U-Tube Viscometer
Certifications/Compliance UKAS, ISO 9000
Volume (Metric) 18 mL
View More Specs
Products 8
Product Code Brand Size Viscosity Range Price Quantity & Availability  
Product Code Brand Size Viscosity Range Price Quantity & Availability  
13161023
View accessory products View Documents
Poulten Selfe™
1619/01
A 0.9 to 3 cSt
232.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13171023
View accessory products View Documents
Poulten Selfe™
1619/02
B 2 to 10 cSt
239.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13101033
View accessory products View Documents
Poulten Selfe™
1619/05
E 60 to 300 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13121033
View accessory products View Documents
Poulten Selfe™
1619/07
G 600 to 3000 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13131033
View accessory products View Documents
Poulten Selfe™
1619/08
H 2000 to 10,000 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13191023
View accessory products View Documents
Poulten Selfe™
1619/04
D 20 to 100 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13181023
View accessory products View Documents
Poulten Selfe™
1619/03
C 6 to 30 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
13111033
View accessory products View Documents
Poulten Selfe™
1619/06
F 200 to 1000 cSt
227.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
Description
Description
  • PSL calibrated viscometers are supplied with UKAS certificates providing an official assurance of the competence of the calibration work carried out. UKAS certificates of calibration, which satisfy the requirements of ISO 9000, are issued by the PSL Calibration Laboratory, which is a National Measurement Accreditation Service accredited laboratory.
  • Type BS/U, IP71.Suitable for transparent liquids. Nominal overall length 300mm. Approx. sample size 13mL to 40mL.
Specifications
Specifications
U-Tube Viscometer
UKAS, ISO 9000
18 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Poulten Selfe™ U-Tube Viscometer