missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBLCP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12099-1-AP-20UL
This item is not returnable.
View return policy
Description
UBLCP1 may specifically dephosphorylate ′Ser-5′ of the heptad repeats YSPTSPS in the C-terminal domain of the largest RNA polymerase II subunit.Specifications
UBLCP1 | |
Polyclonal | |
Unconjugated | |
UBLCP1 | |
CPUB1, UBLCP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134510, 360514, 79560 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5FWT7, Q8BGR9, Q8WVY7 | |
Ublcp1 | |
UBLCP1 Fusion Protein Ag2743 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction