missing translation for 'onlineSavingsMsg'
Learn More

UBLCP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen UBLCP1
Concentration 0.33 mg/mL
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16839613
View Documents
Proteintech
12099-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16829613
View Documents
Proteintech
12099-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

UBLCP1 may specifically dephosphorylate ′Ser-5′ of the heptad repeats YSPTSPS in the C-terminal domain of the largest RNA polymerase II subunit.
Specifications

Specifications

UBLCP1
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Rabbit
Rat, Mouse, Human
Q5FWT7, Q8BGR9, Q8WVY7
134510, 360514, 79560
UBLCP1 Fusion Protein Ag2743
Primary
-20°C
UBLCP1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CPUB1, UBLCP1
Ublcp1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.