Learn More
Retsch Ultra Centrifugal Mill ZM 300
Rotor mills are used for high-speed size reduction of soft to medium-hard as well as temperature-sensitive or fibrous materials.
Brand:  Retsch 20.824.0001
Product Code. 17629295
  • 10360.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The powerful Ultra Centrifugal Mill ZM 300 provides maximum grinding performance combined with ease of use.The variable speed from 6000 to 23,000 rpm allows for gentle, neutral-to-analysis sample preparation in a very short time.
  • Thanks to an integrated temperature monitoring system, reproducibility is guaranteed even for long grinding processes or pulverization of large sample volumes.
  • The wide selection of rotors, ring sieves and cassettes makes the ZM 300 a true allrounder which meets the requirements of a great variety of size reduction tasks.
Specifications
Specifications
Ultra-Centrifugal Mill
900 mL (standard cassette); 1200 mL (volume cassette); 5000 mL / 3000 mL / 500 mL / 250 mL (cyclone)
426 x 452 x 431 mm
Agriculture, biology, chemistry/plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, medicine/pharmaceuticals
1-phase
100/120 V or 200/240 V
452 mm
Not Autoclavable
CE
Reusable
IP20
6000 to 23,000 min-1, free selectable
38 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Retsch Ultra Centrifugal Mill ZM 300 > Ultra-Centrifugal Mill