Learn More
Biochrom Ultrospec 10 Disposable Cell
Ultrospec 10 is a disposable cuvette suited for use in OD600 measurements of cell cultures of bacteria and yeast.
Brand:  Biochrom 80-2004-53
Product Code. 11801772
  • 118.00 € / Pack of 100
Explore more special offers
Description
Description
  • For use with Ultrospec 10
  • Made of methacrylate
  • Min. operating volume is 2.5 mL
Specifications
Specifications
Disposable Cell
OD600
9 V
Plastic
5 to 9
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Biochrom Ultrospec 10 Disposable Cell > Ultrospec 10 Disposable Cell; Cell Density: OD600