Learn More
Hughes Unheated Emergency Safety Shower with Eye/Face Wash
Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit. This ANSI compliant model is ideal for use in indoor applications not subject to the possibility of freezing or overheating.
Brand: Hughes HSHD0018GV
This item is not returnable.
View return policy
Description
- Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit
- ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
- Fitted with foot panel as standard
- Delivers the ANSI recommended 76 liters per minute of potable water from the shower and 12 liters per minute to the eye/face wash
- Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Galvanized Steel | |
Floor | |
Safety Shower |
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower with Eye/Face Wash > Safety Shower; Material: Galvanized Steel
Spot an opportunity for improvement?Share a Content Correction