Learn More
Hughes Unheated Emergency Safety Shower with Eye/Face Wash
Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit. This ANSI compliant model is ideal for use in indoor applications not subject to the possibility of freezing or overheating.
1255.00 € - 1470.00 €
Specifications
Mounting Floor
Product Type Safety Shower
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
17702348
View Documents
Hughes
HSHD0018GV
Galvanized Steel
1255.00 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17712348
View Documents
Hughes
HSHD0018SS
Stainless Steel
1470.00 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
Description
Description
  • Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit
  • ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
  • Fitted with foot panel as standard
  • Delivers the ANSI recommended 76 liters per minute of potable water from the shower and 12 liters per minute to the eye/face wash
  • Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Specifications
Floor
Safety Shower
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower with Eye/Face Wash