missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abeba™ UNI6 1740 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
109.00 € - 113.00 €
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 S2 SRC |
---|---|
Color | White |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15713299
|
Abeba™
1740/35 |
35 |
113.00 €
1 pair |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15723299
|
Abeba™
1740/36 |
36 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15755248
|
Abeba™
1740/37 |
37 |
113.00 €
1 pair |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
15765248
|
Abeba™
1740/38 |
38 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15775248
|
Abeba™
1740/39 |
39 |
109.00 €
1 pair |
Estimated Shipment: 08-10-2025
Log in to see stock. |
||||||
15687668
|
Abeba™
1740/40 |
40 |
109.00 €
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15697668
|
Abeba™
1740/41 |
41 |
109.00 €
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15785248
|
Abeba™
1740/42 |
42 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15757537
|
Abeba™
1740/43 |
43 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15795248
|
Abeba™
1740/44 |
44 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15705258
|
Abeba™
1740/45 |
45 |
109.00 €
1 pair |
Estimated Shipment: 20-10-2025
Log in to see stock. |
||||||
15767537
|
Abeba™
1740/46 |
46 |
109.00 €
1 pair |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
15733299
|
Abeba™
1740/47 |
47 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15743299
|
Abeba™
1740/48 |
48 |
109.00 €
1 pair |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
Description
- CE EN ISO 20345:2011 S2 SRC
- Microfiber
- White
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with elastic band
- **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Polyurethane | |
Steel |
White | |
Slip-resistant | |
1740 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abeba™ UNI6 1740 Shoes