Learn More
ABEBA™ Uni6 Black Occupational Slip-On Shoes O2
ABEBA™ Uni6 Black Occupational Slip-On Shoes O2 made of water and chemical resistant microfiber with a washable, slip resistant PU/TPU outsole.
Brand:  ABEBA™ 36741/42
 View more versions of this product
Product Code. 16388131
  • 102.00 € / 1 pair
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hygiene, comfort
  • Comply to EN ISO 20347 : 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water and chemical resistant microfiber
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
  • Washable at 30°C
Specifications
Specifications
CE EN ISO 20347 :2012, O2, FO, SRC, ESD
Breathable silver ions lining
42
Polyurethane, Thermoplastic Polyurethane
1 Pair
Water
Black
Microfiber
Slip and abrasion resistant, ESD dissipative, shock absorbent
Slip-on shoes
EN ISO 20347 : 2012; DIN EN 61340
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ABEBA™ Uni6 Black Occupational Slip-On Shoes O2 > Size: 42