Learn More
ABEBA™ Uni6 Black Occupational Sneakers O2
ABEBA™ Uni6 Black Occupational Sneakers O2 made of tear and water resistant fabric with slip resistant PU/TPU outsole.
Brand:  ABEBA™ 36728/38
 View more versions of this product
Product Code. 16328091
  • 107.00 € / 1 pair
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, supple, water resistant
  • Comply to EN ISO 20347 : 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Tear and water resistant fabrics
  • Easy fastening with laces
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
Specifications
Specifications
CE EN ISO 20347 :2012, O2, FO, SRC, ESD
Black
Breathable silver ions lining
38
Polyurethane, Thermoplastic Polyurethane
1 Pair
Water
Laces
Stainless steel
Textile
Slip and abrasion resistant, ESD dissipative, shock absorbent
Trainer
EN ISO 20347 : 2012; DIN EN 61340
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ABEBA™ Uni6 Black Occupational Sneakers O2 > Size: 38