missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABEBA™ Uni6 Black Occupational Sneakers O2
ABEBA™ Uni6 Black Occupational Sneakers O2 made of tear and water resistant fabric with slip resistant PU/TPU outsole.
107.00 € - 111.00 €
Specifications
Certifications/Compliance | CE EN ISO 20347 :2012, O2, FO, SRC, ESD |
---|---|
Closure Type | Laces |
Color | Black |
Eyelets Material | Stainless steel |
Liner Material | Breathable silver ions lining |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16398081
|
ABEBA™
36728/35 |
35 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16308091
|
ABEBA™
36728/36 |
36 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16318091
|
ABEBA™
36728/37 |
37 |
111.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16328091
|
ABEBA™
36728/38 |
38 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16338091
|
ABEBA™
36728/39 |
39 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16348091
|
ABEBA™
36728/40 |
40 |
110.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16358091
|
ABEBA™
36728/41 |
41 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16368091
|
ABEBA™
36728/42 |
42 |
110.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16378091
|
ABEBA™
36728/43 |
43 |
111.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16388091
|
ABEBA™
36728/44 |
44 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16398091
|
ABEBA™
36728/45 |
45 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16308101
|
ABEBA™
36728/46 |
46 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16318101
|
ABEBA™
36728/47 |
47 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
16328101
|
ABEBA™
36728/48 |
48 |
107.00 €
1 pair |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
- Lightweight, supple, water resistant
- Comply to EN ISO 20347 : 2012, O2, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- Good cushioning
- Energy absorption in heel area
- Tear and water resistant fabrics
- Easy fastening with laces
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- Removable Active Comfort insole
Specifications
CE EN ISO 20347 :2012, O2, FO, SRC, ESD | |
Black | |
Breathable silver ions lining | |
Slip and abrasion resistant, ESD dissipative, shock absorbent | |
Trainer | |
EN ISO 20347 : 2012; DIN EN 61340 | |
Yes |
Laces | |
Stainless steel | |
Textile | |
Polyurethane, Thermoplastic Polyurethane | |
1 Pair | |
Water |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ABEBA™ Uni6 Black Occupational Sneakers O2