Learn More
ABEBA™ Uni6 Black Safety Trainers S3 ESD
Breathable, water repellent trainers made of knitted fabric, slip resistant PU/TPU outsole, steel toe cap, textile antipuncture
Brand:  ABEBA™ 31676/42
 View more versions of this product
Product Code. 17101185
  • 123.00 € / 1 pair
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made for safety and comfort
  • Comply to EN ISO 20345: 2012, S3, SRC
  • ESD dissipative according EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Specifications
Black
42
ESD according 61340 class climatic 1, EN ISO 20345: 2012 S3 SRC
Trainers
S3 SRC ESD
Knitted Fabric
Steel
Unisex
Water Repellent