Learn More
Thermo Scientific™ Universal Sample Holders for Thermo Scientific Spectronic™ IR100 FT-IR Spectrometer
Brand: Thermo Scientific™ 0016-225
This item is not returnable.
View return policy
Description
Universal Sample Holders. Available in two sizes.
- Mounts for salt plates, pellets, or films
- Fit in standard slide-card mounts
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Spectronic IR100 FT-IR spectrometer | |
Universal Sample Holder | |
25 mm | |
For positioning of KBr pellets, thin films, or IR windows for transmission analyses |