Learn More
Thermo Scientific™ Universal Sample Holders for Thermo Scientific Spectronic™ IR100 FT-IR Spectrometer
436.00 €
Specifications
For Use With (Equipment) Spectronic IR100 FT-IR spectrometer
Type Universal Sample Holder
For Use With (Application) For positioning of KBr pellets, thin films, or IR windows for transmission analyses
View More Specs
Products 1
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
15890727
View Documents
Thermo Scientific™
0016-225
25 mm
436.00 €
Each
Description
Description
Universal Sample Holders. Available in two sizes.
  • Mounts for salt plates, pellets, or films
  • Fit in standard slide-card mounts
Specifications
Specifications
Spectronic IR100 FT-IR spectrometer
Universal Sample Holder
For positioning of KBr pellets, thin films, or IR windows for transmission analyses