Learn More
asecos™ Universal Sorbents Sock
The Universal Sorbents Sock is ideal for the safe absorption of oils, water, coolants and non-aggressive chemicals.
Brand:  asecos™ 11347
 View more versions of this product
Product Code. 17896416
  • 124.00 € / Pack of 6
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibers, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Specifications
White
Universal Sorbent Sock
3 m
75.00 mm
6 kg
Heavy-duty Cloth
9.84 ft.
2.95 in.
13.23 lb.