missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Peek Low-Pressure Union Assembly

Make solid connections in low-pressure applications

Brand:  Kinesis™ P-702

 View more versions of this product

Product Code. 13020390

  • 53.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Make solid connections in low-pressure applications

  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • Max pressure of 1000 psi (69 bar)
  • 0.030 in. thru-hole diameter
  • 0.41 μL swept volume
Specifications

Specifications

Union
80°C
Natural
1000 psi
PEEK
With 2 P-200/P-235, 1/16 in., 1/4-28, 0.020 in. (0.5 mm) THRU
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers