missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Brand:  Kinesis™ 02010-84

 View more versions of this product

Product Code. 13020850

  • 16.15 € / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Specifications

Tubing
1/16 in.
0.5 mm
20 cm
15,800 psi (1089 bar)
Yellow
0.020 in.
8 in.
Stainless Steel (304)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers