Learn More
Kinesis™ Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID
Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses
9.45 € - 25.35 €
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Material | Price | Quantity & Availability | |||
10752527
|
Kinesis™
U-128 |
0.175 mm | 20 cm | Stainless Steel (316) | N/A | ||||
13000910
|
Kinesis™
02011-36 |
1.0 mm | 5 cm | Stainless Steel (316) |
14.05 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
13000850
|
Kinesis™
02010-82 |
0.5 mm | 5 cm | Stainless Steel (304) |
9.45 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
13020850
|
Kinesis™
02010-84 |
0.5 mm | 20 cm | Stainless Steel (304) |
16.15 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
13080850
|
Kinesis™
02010-90 |
0.25 mm | 5 cm | Stainless Steel (316) |
9.95 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
13050920
|
Kinesis™
02011-51 |
0.125 mm | 10 cm | Stainless Steel (316) |
13.30 €
Each |
In Stock
Log in to see stock. |
|||
13070920
|
Kinesis™
02011-53 |
0.125 mm | 30 cm | Stainless Steel (316) |
25.35 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
Tubing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.