Learn More
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
49.10 € - 477.00 €
Chemical Identifiers
CAS | 124-43-6, 7722-88-5 |
---|---|
Molecular Formula | CH6N2O3 |
Molecular Weight (g/mol) | 94.07 |
InChI Key | AQLJVWUFPCUVLO-UHFFFAOYSA-N |
Synonym | urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid |
PubChem CID | 31294 |
ChEBI | CHEBI:75178 |
IUPAC Name | hydrogen peroxide;urea |
SMILES | C(=O)(N)N.OO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10420321
|
Thermo Scientific Acros
241022500 |
250 g | Glass bottle |
49.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266620
|
Thermo Scientific Acros
241020010 |
1 kg | Glass bottle |
129.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924381
|
Thermo Scientific Acros
241020051 |
5 kg | Plastic drum |
477.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
124-43-6, 7722-88-5 | |
94.07 | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
CHEBI:75178 | |
C(=O)(N)N.OO |
CH6N2O3 | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
31294 | |
hydrogen peroxide;urea |
Specifications
124-43-6, 7722-88-5 | |
White | |
CO(NH2)2·H2O2 | |
15,1784 | |
Solubility in water: 500g/L (20°C). Other solubilities: alcohol and ether decomposes the product to h2o2, and urea, soluble in ethylene glycol | |
C(=O)(N)N.OO | |
1% sodium pyrophosphate | |
31294 | |
94.07 | |
Tablets |
75.0°C to 85.0°C | |
CH6N2O3 | |
16,379; 17,401 | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
hydrogen peroxide;urea | |
94.07 | |
CHEBI:75178 | |
≥33.5% (H2O2) | |
Urea hydrogen peroxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-701-4
- Urea hydrogen peroxide (1 g tablets), stabilized, contains 35 wt% H2O2
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only