missing translation for 'onlineSavingsMsg'
Learn More
Uric acid, 99%, Thermo Scientific Chemicals Product Code.: 11410973

Uric acid, 99%, Thermo Scientific Chemicals

Product Code. 11410973
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11410973

Brand: Thermo Scientific Alfa Aesar A13346.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Uric acid acts as a strong reducing agent (electron donors) and potent antioxidant. It is used as a precursor in cell culture systems and in cosmetics. Further, it is useful for serum-free medium supplement. In addition to this, it is used as a reagent in the determination of uricase and tungstate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uric acid acts as a strong reducing agent (electron donors) and potent antioxidant. It is used as a precursor in cell culture systems and in cosmetics. Further, it is useful for serum-free medium supplement. In addition to this, it is used as a reagent in the determination of uricase and tungstate.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water. Insoluble in ether and alcohol.

Notes
Stable. Incompatible with other materials such as acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-93-2
Molecular Formula C5H4N4O3
Molecular Weight (g/mol) 168.112
MDL Number MFCD00005712
InChI Key LEHOTFFKMJEONL-UHFFFAOYSA-N
Synonym uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydroShow MoreShow Less
PubChem CID 1175
ChEBI CHEBI:17775
IUPAC Name 7,9-dihydro-3H-purine-2,6,8-trione
SMILES C12=C(NC(=O)N1)NC(=O)NC2=O

Specifications

Melting Point >300°C
Density 1.89
Quantity 25 g
Beilstein 156158
Merck Index 14,9875
Solubility Information Soluble in 1M sodium hydroxide solution. Slightly soluble in water. Insoluble in ether and alcohol.
Formula Weight 168.11
Percent Purity 99%
Chemical Name or Material Uric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP06L022 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberA13346.22,A13346.14,A13346.36
Lot NumberW27K015 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberA13346.22,A13346.14,A13346.36
Lot NumberS12K031 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberA13346.22,A13346.14,A13346.36
Lot NumberR09K019 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog NumberA13346.22,A13346.14,A13346.36
Lot NumberZ08J013 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog NumberA13346.22,A13346.14,A13346.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Uric acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.