missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Lamp

Single wavelength UV lamp with filter that is an ideal solution for research and enumeration of E. coli.

Brand:  Vilber Lourmat™ 3111-2153-1

 View more versions of this product

Product Code. 11746668

  • 943.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Single wavelength UV lamp with filter
  • 6 W UV lamp
  • Rapid visualization of CCM plates requiring UV revelation
  • Complete with UV tube and filter
  • Research and enumeration of E. coli
  • UV lamp 254nm
Specifications

Specifications

30 W
140 mm
505 mm
1 Ea.
365 nm
117 mm
2300 mW/cm2
UV Lamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.