missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Lamp

Single wavelength UV lamp with filter that is an ideal solution for research and enumeration of E. coli.

Brand:  Vilber Lourmat™ 3111 2155 1

 View more versions of this product

Product Code. 11796658

  • 945.00 € / Each
Estimated Shipment: 19-09-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Single wavelength UV lamp with filter
  • 6 W UV lamp
  • Rapid visualization of CCM plates requiring UV revelation
  • Complete with UV tube and filter
  • Research and enumeration of E. coli
  • UV lamp 254nm
Specifications

Specifications

UV Lamp
365/254 nm
117 mm
1350/1100 mW/cm2
30 W
140 mm
505 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers