Learn More
uvex Profapren CF33 Chemical Protecion Gloves
High-quality unsupported chloroprene safety gloves for protection against a broad spectrum of chemicals
11.25 € - 14.05 €
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15336922
View Documents
uvex
6011901
7
11.25 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
15346922
View Documents
Uvex™
6011902
8
13.45 €
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
15356922
View Documents
uvex
6011903
9
11.25 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
15316922
View Documents
uvex
6011904
10
14.05 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
15326922
View Documents
Uvex™
6011905
11
13.45 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
Description
Description
  • Silicone-free safety gloves with excellent properties for protection against chemical and mechanical risks
  • Good combination of flexibility and strength
  • Resistant to many chemicals and solvents
  • EN 388:2016 (3131X)
  • EN ISO 374-1: 2016 / Type A (AKLMNO)
  • Videos