Learn More
UVP Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Brand:  UVP 34-0068-02
 View more versions of this product
Product Code. 12813858
  • 96.60 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
40 W
365 nm
Model XX-40 UV bench lamp
1 Ea.