missing translation for 'onlineSavingsMsg'
Learn More

UVP Replacement UV Light Tubes

Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators

27.40 € - 96.60 €


Products 4
Product Code Brand For Use With (Equipment) Wattage Wavelength Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Wattage Wavelength Price Quantity & Availability  
12842989
View Documents
UVP
34-0044-01
EL-series UV lamp, 3UV™ lamp, handheld UV lamp, mid size UV viewing cabinets, benchtop UV transilluminators 6 W 302 nm
62.90 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
12813858
View Documents
UVP
34-0068-02
Model XX-40 UV bench lamp 40 W 365 nm
96.60 €
Each
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
 
12893848
View Documents
UVP
34-0068-01
Model XX-20BLB UV bench lamp 20 W 365 nm
60.30 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
12883918
View Documents
UVP
34-0009-02
Model XX-15L UV bench lamp, ChromaDoc-It™ imaging system and Chromato-Vue™ C-70G, C-71, C-75 large UV viewing cabinets 15 W 365 nm
27.40 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
Description

Description

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.