missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva VacuCap™ 90 PF Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva
208.05 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications

Specifications
Supor (Hydrophilic PES) | |
Sterile | |
60 mm |
0.8/0.2 μm | |
30 cm2 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva VacuCap™ 90 PF Vacuum Filtration Devices > 60 mm
Spot an opportunity for improvement?Share a Content Correction