Learn More
Welch Ilmvac™ Vacuum Pump Oil
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
Brand:  Welch Ilmvac™ 800124
 View more versions of this product
Product Code. 10420853
  • 319.00 € / 20 L
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Generic mineral oil
  • Use in most one and two stage rotary vane pumps
  • 20L drum
  • Density: 0.888g/mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Pump Oil
10 - 5 mbar
118 cSt viscosity at 40°C
20 L