Learn More
Welch Ilmvac™ Vacuum Pump Oil
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
137.00 € - 319.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10420853
|
Welch Ilmvac™
800124 |
20 L |
319.00 €
20 L |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
10777544
|
Welch Ilmvac™
800120-01 |
5 L |
137.00 €
5 L |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Description
Generic mineral oil for use in most one and two stage rotary vane pumps.Specifications
Vacuum Pump Oil |