Learn More
Thermo Scientific™ Valco Injector, Model C6W
Valco continues to offer a diverse line in terms of number of ports, fitting sizes, and materials of construction.
Brand:  Thermo Scientific™ EPC6W
2305.44 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11553970
  • 2316.97 € / Each
Explore more special offers
Specifications
Microelectric
EPC6 W
Injector
20 μL Loop
Six 0.016 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Valco Injector, Model C6W > Model EPC6W injector; Microelectric actuator