missing translation for 'onlineSavingsMsg'
Learn More

Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Form: approx. 3.87g/50x100mm

157.79 € - 427.13 €

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311319
SDS View Documents
Thermo Scientific Alfa Aesar
013783.FY
50 x 100 mm
157.79 €
Each
Please to purchase this item. Need a web account? Register with us today!
11321319
SDS View Documents
Thermo Scientific Alfa Aesar
013783.GK
100 x 200 mm
427.13 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-62-2
50.94
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]

Specifications

Silver to Gray
Silver Gray foil
99.8% (Metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
23990
Odorless
Vanadium
7440-62-2
6.11g/cm3
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
50.94
CHEBI:35170
Foil
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-171-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.