missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chl, Thermo Scientific Chemicals

99.20 € - 537.00 €

Chemical Identifiers

CAS 19559-06-9, 75-09-2
Molecular Formula VCl3·C12H24O3
Molecular Weight (g/mol) 373.62
InChI Key WARDLUZTYLPMGJ-UHFFFAOYSA-K
Synonym trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex
PubChem CID 71311352
IUPAC Name oxolane;trichlorovanadium
SMILES C1CCOC1.Cl[V](Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11986171
View Documents Promotion Details
Thermo Scientific Acros
433661000
100 mL
99.20 €
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11996171
View Documents Promotion Details
Thermo Scientific Acros
433668000
800 mL
537.00 €
800mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19559-06-9, 75-09-2
373.62
trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex
oxolane;trichlorovanadium
VCl3·C12H24O3
WARDLUZTYLPMGJ-UHFFFAOYSA-K
71311352
C1CCOC1.Cl[V](Cl)Cl

Specifications

19559-06-9, 75-09-2
Solution
VCl·3THF
trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex
C1CCOC1.Cl[V](Cl)Cl
373.62
373.62
AcroSeal™ Glass bottle
1.3120g/mL
Red
VCl3·C12H24O3
−8°C
WARDLUZTYLPMGJ-UHFFFAOYSA-K
oxolane;trichlorovanadium
71311352
0.40 to 0.60M
1.312
Vanadium(III) chloride-tetrahydrofuran complex (1:3)
Videos
Safety and Handling

Safety and Handling

GHS P Statement Highly flammable liquid and vapour. Suspected of causing cancer. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause drowsiness or dizziness. May cause damage to organs

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

Product Identifier
  • Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only