missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment

35.38 € - 196.98 €

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374919
SDS View Documents
Thermo Scientific Alfa Aesar
081119.09
10 g
35.38 €
10g
Please to purchase this item. Need a web account? Register with us today!
11384919
SDS View Documents
Thermo Scientific Alfa Aesar
081119.18
50 g
82.11 €
50g
Please to purchase this item. Need a web account? Register with us today!
11394919
SDS View Documents
Thermo Scientific Alfa Aesar
081119.30
250 g
196.98 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3153-26-2
265.16
JFHJZWAQYMGNBE-SUKNRPLKSA-L
131674261
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14O5V
MFCD00000032
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate

Specifications

140°C (13mmHg)
3153-26-2
C10H14O5V
UN3285
Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
265.16
265.16
Vanadium (IV) oxide bis (2,4-pentanedionate)
256°C to 259°C
Powder
MFCD00000032
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
JFHJZWAQYMGNBE-SUKNRPLKSA-L
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
131674261
1.4 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

missing translation for 'einecsNumber' : 221-590-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.